Learn More
trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile, TRC
Specifications
Specifications
| Chemical Name or Material | trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile |
| CAS | 300364-84-5 |
| Molecular Formula | C17 H18 N2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C17H18N2/c1-13(10-15(11-18)12-19)9-14-5-7-16(8-6-14)17(2,3)4/h5-10H,1-4H3/b13-9+ |
| SMILES | C\C(=C/c1ccc(cc1)C(C)(C)C)\C=C(C#N)C#N |
| IUPAC Name | 2-[(E)-3-(4-tert-butylphenyl)-2-methyl-prop-2-enylidene]propanedinitrile |
| Molecular Weight (g/mol) | 250.34 |
| Formula Weight | 250.147 |