Learn More
3M™ Tracer™ Replacement Pod
Product Code. 12652615
Click to view available options
Color:
Blue
Dark Blue
Unit Size:
50 pairs
Pack of 50
Product Code. 12652615
Brand: 3M™ 7000103758
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The Tracer™ Replacement Pod is designed for insertion into the ear canal to help reduce exposure to hazardous levels of noise up to 20 dB.
  • Reduces noise levels up to 20 dB SNR in accordance with EN 352-2:2020.
  • Patented tri-flange design in pre-moulded material makes them a fit for most ear canals.
  • Firm, long stem for easy insertion and removal.
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Quantity 20/Pk.
Product Type Earplug