Learn More
Thermo Scientific™ TracePLOT TG-BOND Alumina KCl Metal
Product Code. 13448968 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
5 m
Unit Size:
Each
Product Code. 13448968
Brand: Thermo Scientific™ 26M026170
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For consistent, reproducible analysis of nonpolar and polar compounds, choose the polar PLOT column with essentially no particle release.
  • For linear and quantitative of polar unsaturated hydrocarbonds
  • Available with either Na2SO4 or KCl deactivation
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 0.53 mm
Stationary Phase TG-BOND Alumina KCl
Film Thickness 10 μm
Length (Metric) 5 m