Learn More
Thermo Scientific™ TraceGOLD TG-VRX GC Columns
Product Code. 11561164 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
105 m
20 m
30 m
40 m
60 m
75 m
Diameter (Metric):
0.18 mm
0.25 mm
0.32 mm
0.53 mm
Unit Size:
Each
Product Code. 11561164
Brand: Thermo Scientific™ 260814080
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Thermo Scientific™ TraceGOLD TG-VRX GC Columns provide good resolution and fast analysis times for volatile compounds.
Thermo Scientific™ TraceGOLD TG-VRX GC Columns provide good resolution and fast analysis times for volatile compounds.
  • Proprietary stationary phase
  • Optimized dimensions for analysis of volatile organic pollutants
  • Ideally suited for US EPA Methods 502.2, 524.2, 8021, and 8260
  • Similar to DB-VRX, Rtx-VRX
Recommended for:
  • Volatile organic pollutants
  • Aromatics
TRUSTED_SUSTAINABILITY
Specifications
Max. Temperature 260°C
Diameter (Metric) 0.53 mm
Diameter (Metric) Inner 0.53 mm
Stationary Phase TraceGOLD™ TG-VRX
For Use With (Application) GC/MS for Volatile Organic Pollutants, GC/MS for US EPA Method 8021 Compounds
Film Thickness 3 μm
Length (Metric) 60 m
Polarity Non-Polar
Description 60m x 0.53mm x 3.00um