Learn More
Thermo Scientific™ TraceGOLD TG-VMS GC Columns
Product Code. 11501164 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
20 m
30 m
40 m
60 m
75 m
Diameter (Metric):
0.18 mm
0.25 mm
0.32 mm
0.53 mm
Unit Size:
Each
Product Code. 11501164
Brand: Thermo Scientific™ 260804960
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Thermo Scientific™ TraceGOLD TG-VMS GC Columns provide good resolution and fast analysis times for volatile compounds.
Thermo Scientific™ TraceGOLD TG-VMS GC Columns provide good resolution and fast analysis times for volatile compounds.
  • Proprietary stationary phase
  • Optimized dimensions for analysis of volatile organic pollutants
  • Ideally suited for US EPA Method 8260B
  • Similar to Rtx-VMS
Recommended for:
  • Volatile organic pollutants
TRUSTED_SUSTAINABILITY
Specifications
Max. Temperature 260°C
Diameter (Metric) 0.18 mm
Diameter (Metric) Inner 0.18 mm
Stationary Phase TraceGOLD™ TG-VMS
For Use With (Application) GC/MS for Volatile Organic Pollutants, GC/MS for US EPA Method 8260B Compounds
Film Thickness 1 μm
Length (Metric) 40 m
Polarity Non-Polar