Learn More
Thermo Scientific™ TraceGOLD TG-5SilMS GC Columns
Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
12 m
15 m
20 m
30 m
40 m
60 m
Diameter (Metric) Inner:
0.18 mm
0.2 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.18 μm
0.25 μm
0.33 μm
0.5 μm
1 μm
1.5 μm
Unit Size:
Each
Description
Thermo Scientific™ TraceGOLD™ TG-5SilMS GC Columns incorporate phenyl groups in the polymer backbone for improved thermal stability, reduced bleed and reduced susceptibility to oxidation.
- Low polarity, silarylene phase with selectivity comparable to 5% diphenyl/95% dimethyl polysiloxane
- Designed for low bleed and outstanding inertness
- Similar to DB-5MS, VF-5ms, CP-Sil 8 Low-Bleed/MS, Rxi-5Sil MS, BPX5, ZB-5ms, Optima-5MS, SLB-5
Recommended for:
- GC/MS applications using ion-trap systems
- Polycyclic aromatics
- Hydrocarbons including chlorinated hydrocarbons
- Phthalates
- Phenols
- Amines
- Organophosphates
Specifications
Specifications
| Max. Temperature | 330/350°C |
| Diameter (Metric) | 0.2 mm |
| Diameter (Metric) Inner | 0.2 mm |
| Stationary Phase | TraceGOLD™ TG-5 SilMS |
| USP Type | G27, G36 |
| For Use With (Application) | GC/MS Applications Using Ion-Trap Systems, GC/MS for Polycyclic Aromatics, Phthalates, Phenols, Amines, and Organophosphates, GC/MS for Hydrocarbons Including Chlorinated Hydrocarbons |
| Film Thickness | 0.33 μm |
| Length (Metric) | 12 m |
| Polarity | Low-Polar |
| Description | TG-5SilMS 12m x 0.2mm x 0.33µm |