Learn More
Thermo Scientific™ TraceGOLD™ TG-5MT Metal GC Columns
Product Code. 11860473 Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Length (Metric):
15 m
30 m
60 m
Diameter (Metric):
0.25 mm
0.53 mm
Film Thickness:
0.1 μm
0.25 μm
0.5 μm
1 μm
1.5 μm
3 μm
missing translation for 'unitSize'
Each
Product Code. 11860473
missing translation for 'mfr': Thermo Scientific™ 26M981420
Only null left
Add to basket
Add to basket
Give greater stability and inertness at elevated temperatures and when working with highly active compounds with Thermo Scientific™ TraceGOLD™ TG-5MT Metal GC Columns.
TG-5MT Metal GC Columns provide greater robustness under increased stress.
  • Can be used at elevated temperature without risk of bleed or breakdown
  • Excellent inertness when working with highly active compounds
  • Robust under stress
TRUSTED_SUSTAINABILITY
Specifications
Max. Temperature 330/350°C (Standard Columns), 430°C (Metal Columns)
Diameter (Metric) 0.25 mm
Diameter (Metric) Inner 0.25 mm
Stationary Phase TraceGOLD™ TG-5 MT
USP Type G27, G36
For Use With (Application) Solvent Impurities Analysis, GC/MS for Residual Solvents and Solvent Impurities, GC/MS for Drugs of Abuse, Pesticides, PCB Cogeners, and Aroclor Mixes
Particle Size 0.25 μm
Temperature Range -60°C to +430°C
Film Thickness 0.25 μm
Length (Metric) 30 m
Polarity Low-Polar