Learn More
Thermo Scientific™ TraceGOLD™ TG-5MS GC Columns
Product Code. 10584685 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 m
12 m
15 m
20 m
30 m
40 m
60 m
Diameter (Metric) Inner:
0.1 mm
0.15 mm
0.18 mm
0.2 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.15 μm
0.18 μm
0.25 μm
0.33 μm
0.5 μm
1 μm
1.5 μm
5 μm
Unit Size:
Each
32 product options available for selection
Product selection table with 32 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Inner Film Thickness Unit Size
10584685 30 m 0.25 mm 0.25 μm Each
10723297 10 m 0.1 mm 0.1 μm Each
15207488 12 m 0.2 mm 0.33 μm Each
13408018 40 m 0.15 mm 0.15 μm Each
13488008 20 m 0.15 mm 0.15 μm Each
15581790 30 m 0.53 mm 5 μm Each
10649105 15 m 0.25 mm 0.25 μm Each
10066504 15 m 0.32 mm 0.25 μm Each
10137334 15 m 0.53 mm 0.25 μm Each
10086644 30 m 0.32 mm 0.25 μm Each
10416954 30 m 0.53 mm 0.25 μm Each
10659295 60 m 0.25 mm 0.25 μm Each
10289754 60 m 0.32 mm 0.25 μm Each
10085884 15 m 0.25 mm 0.5 μm Each
10086314 15 m 0.32 mm 0.5 μm Each
10096314 15 m 0.53 mm 0.5 μm Each
10408874 30 m 0.25 mm 0.5 μm Each
10137904 30 m 0.32 mm 0.5 μm Each
10457914 30 m 0.53 mm 0.5 μm Each
10230005 60 m 0.25 mm 0.5 μm Each
10056034 60 m 0.32 mm 0.5 μm Each
10584105 15 m 0.32 mm 1 μm Each
10594105 15 m 0.53 mm 1 μm Each
10650056 30 m 0.25 mm 1 μm Each
10681206 30 m 0.32 mm 1 μm Each
10744257 30 m 0.53 mm 1 μm Each
10576185 60 m 0.25 mm 1 μm Each
10586185 60 m 0.32 mm 1 μm Each
10753117 15 m 0.53 mm 1.5 μm Each
10371025 30 m 0.53 mm 1.5 μm Each
12586299 60 m 0.53 mm 5 μm Each
11531174 20 m 0.18 mm 0.18 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 32 options available.
32 options
Product Code. 10584685 Supplier Thermo Scientific™ Supplier No. 260981420
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
Estimated Shipment: 02-10-2026
Employ the most widely used MS phase in gas chromatography with the 5% phenyl phase Thermo Scientific™ TraceGOLD™ TG-5MS GC Column.
Employ the most widely used MS phase in gas chromatography with the 5% phenyl phase TraceGOLD TG-5MS GC Column.
  • Low polarity phase, 5% diphenyl/95% dimethyl polysiloxane
  • Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity
  • Exceptional inertness ideal for analysis of active compounds
  • Equivalent to USP G27 phase
  • Similar to Rxi™-5ms, DB-5, HP-5, HP-5ms, Ultra-2, SPB-5, Equity-5, CP-Sil 8
Recommended for:
  • Semivolatiles
  • Phenols
  • Amines
  • Residual solvents and solvent impurities
  • Drugs of abuse
  • Pesticides
  • PCB cogeners
  • Aroclor mixes
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Active Compounds Analysis, GC/MS for Analysis of Active Compounds
Particle Size 0.25 μm
Film Thickness 0.25 μm
Length (Metric) 30 m
Polarity Low-Polar
Description 0.25 um Thickness; 0.25 mm ID; 30 m Length
Diameter (Metric) 0.25 mm
USP Type G27, G36
Stationary Phase TraceGOLD™ TG-5 MS
Diameter (Metric) Inner 0.25 mm
Product Line TraceGOLD
Max. Temperature (Metric) 330/350°C