Learn More
Thermo Scientific™ TraceGOLD TG-35MS GC Columns
Product Code. 10016174 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
15 m
30 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 μm
0.5 μm
1 μm
1.5 μm
3 μm
Unit Size:
Each
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Inner Film Thickness Unit Size
10016174 30 m 0.25 mm 0.25 μm Each
10505635 15 m 0.25 mm 0.25 μm Each
10534115 15 m 0.32 mm 0.25 μm Each
10167704 30 m 0.32 mm 0.25 μm Each
10733877 15 m 0.25 mm 0.5 μm Each
10639295 15 m 0.32 mm 0.5 μm Each
10545815 15 m 0.53 mm 0.5 μm Each
10371115 30 m 0.25 mm 0.5 μm Each
10279754 30 m 0.32 mm 0.5 μm Each
10781957 30 m 0.53 mm 0.5 μm Each
10621016 15 m 0.25 mm 1 μm Each
10219804 15 m 0.53 mm 1 μm Each
10629675 30 m 0.25 mm 1 μm Each
10229084 30 m 0.32 mm 1 μm Each
10713297 30 m 0.53 mm 1 μm Each
10791957 15 m 0.53 mm 1.5 μm Each
10534875 30 m 0.53 mm 1.5 μm Each
10381345 30 m 0.53 mm 3 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Product Code. 10016174 Supplier Thermo Scientific™ Supplier No. 260941420
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
Estimated Shipment: 30-09-2026
Use these columns for a higher phenyl content that provides useful elution order and retention time changes.
Thermo Scientific™ TraceGOLD TG-35MS Columns offers a higher phenyl content providing useful elution order and retention time changes.
  • Mid-polarity phase, 35% diphenyl/65% dimethyl polysiloxane
  • Equivalent to USP G42 phase
  • Similar to Rtx™-35, BD-35, HP-35, SPB-35, SPB-608
Recommended for:
  • Organochlorine pesticides and herbicides
  • Pharmaceuticals
  • PCB cogeners
  • Aroclor mixes
  • Sterols
  • Rosin acids
  • Phthalate esters
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Pharmaceuticals Analysis, GC/MS for Pharmaceuticals, GC/MS for PCB Cogeners, GC/MS for Aroclor Mixes, Sterols, Rosin Acids, and Phthalate Esters
Film Thickness 0.25 μm
Length (Metric) 30 m
Polarity Mid-Polar
Description 0.25 um Thickness; 0.25 mm ID; 30 m Length
Diameter (Metric) 0.25 mm
USP Type G42
Stationary Phase TraceGOLD™ TG-35 MS
Diameter (Metric) Inner 0.25 mm
Product Line TraceGOLD
Max. Temperature (Metric) 300/320°C