Learn More
Thermo Scientific™ TraceGOLD TG-200MS GC Columns
Product Code. 13488018 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
20 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
13488018 20 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13488018 Supplier Thermo Scientific™ Supplier No. 260842760
Only null left
Add to basket
Add to basket
Exceptionally inert mid-polarity columns with selectivity and elution order optimized for difficult separations
  • Polar phase, trifluoropropylmethyl polysiloxane solid phase resolves compounds that phenyl and cyano phases cannot
  • Outstanding thermal stability and low bleed
  • Suitable for use with sensitive detectors including ECDs, NPDs and MS
  • Confirmation column in combination with another GC column
  • Equivalent to USP G6 phase
  • Similar to Rtx™-200MS, DB-200, DB-210
TRUSTED_SUSTAINABILITY
Specifications
Max. Temperature -20°C to +320/340°C
Length (English) Capillary 65.61 ft.
Type GC Capillary Column
Diameter (Metric) Inner 0.15 mm
For Use With (Application) Silica Analysis
Film Thickness 0.15 μm
Length (Metric) 20 m