Learn More
Thermo Scientific™ TraceGOLD™ TG-17SilMS GC Columns
Product Code. 15287478 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 m
20 m
30 m
60 m
Diameter (Metric) Inner:
0.15 mm
0.18 mm
0.25 mm
0.32 mm
Film Thickness:
0.15 μm
0.18 μm
0.25 μm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Inner Film Thickness Unit Size
15287478 30 m 0.25 mm 0.25 μm Each
13498018 10 m 0.15 mm 0.15 μm Each
15254533 60 m 0.25 mm 0.25 μm Each
15214533 20 m 0.18 mm 0.18 μm Each
15264533 30 m 0.32 mm 0.25 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15287478 Supplier Thermo Scientific™ Supplier No. 260721420
Estimated Shipment: 11-12-2026
Optimize separation when analyzing active environmental compounds with this low-bleed GC column with superior inertness.
Optimize separation when analyzing active environmental compounds with the Thermo Scientific™ TraceGOLD™ TG-17SilMS GC Column. The column has the stability and performance profile ideal for separation of active environmental compounds such as polycyclic aromatic hydrocarbons (PAHs). Superior inertness ensures excellent peak shape and sensitivity.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Max. Temperature 340/360°C
For Use With (Application) Phthalate Esters Analysis, GC/MS for Pesticides and Herbicides, GC/MS for Phthalate Esters, GC/MS for Triglycerides
Length (Metric) 30 m
Diameter (Metric) Inner 0.25 mm
USP Type G3
Product Line TraceGOLD
Film Thickness 0.25 μm