Learn More
Fisherbrand™ TraceableOne™ Single-Use USB Temperature Data Logger
Product Code. 17357619 Shop All Cole-Parmer Products
Click to view available options
Temperature Range (Metric):
-20°C to -15°C
2°C to 8°C
Data Memory:
16,000 Readings, 10 Day, 1 min. Interval
16,000 Readings, 30 Day, 3 min. Interval
16,000 Readings, 60 Day, 6 min. Interval
16,000 Readings, 90 Day, 10 min. Interval
Unit Size:
Pack of 40
Product Code. 17357619
Brand: Fisherbrand™ 1800404
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The TraceableOne™ Single-Use USB Temperature Data Logger automatically generates PDF data report via USB port.
  • LED alarm indicator for visual status indication
  • Time delay of 30 minutes before logging begins for enhanced logging accuracy
  • Protective plastic pouch maintains waterproof IP67 rating
  • Bar code for in-house scanning
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Specifications
Data Memory 16,000 Readings, 30 Day, 3 min. Interval
Temperature Range (Metric) 2°C to 8°C