Learn More
Fisherbrand™ TraceableOne™ 40 Pack Single-User Datalogger
Product Code. 16019511 Shop All Fisherbrand Products
Click to view available options
Alarm Types:
Preprogrammed -20°C to -15°C
Preprogrammed 2°C to 8°C
Data Memory:
10 Day, 1 min. Interval
30 Day, 3 min. Interval
60 Day, 6 min. Interval
90 Days, 10 min. Interval
Unit Size:
Pack of 40
Product Code. 16019511
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use logger right out of the box to track environmental conditions of reagents, clinical samples, vaccines, pharmaceuticals and food items during transit. Fisherbrand™ TraceableOne™ 40 Pack Single-User Datalogger allows for monitoring materials every step of the way.
  • Logger generates PDF report
  • Download directly to PC
  • No Software required
  • Compact
  • Single-use
  • Flashing Red and Green LED's indicate if logger is recording data, has stopped recording and if an alarm has been triggered
  • Press start to begin recording crucial temperature data, 30 minute time delay before logging begins
Specifications
Type Single-use Datalogger
Data Memory 10 Day, 1 min. Interval
Calibration Calibrated
Alarm Types Preprogrammed -20°C to -15°C
Software Type No software required, download data as PDF report
Description 40/Pk
Includes Batch-calibrated Traceable certificate
IP Rating IP67
For Use With (Application) Refrigerated transportation samples
Accuracy (Temperature) ±0.5°C from -20° to 40°C, otherwise ±1°C
Temperature Resolution 0.01°
Temperature Range (Metric) -30°C to +70°C
Length (Metric) 8.89 cm
Electrical Requirements Battery-Powered