Learn More
Alla France™ Traceable Mineral Oil Hydrometer
Click to view available options
Specific Gravity Range:
29 to 41° API
9 to 21° API
Unit Size:
Each
Specifications
Specifications
| Quantity | 1Each |
| Type | Traceable Mineral Oil Hydrometer |
| Accuracy | ±0.1°API |
| Length (Metric) | 335 mm |
| Baume Range | 29 to 41°API |
| Specific Gravity Range | 29 to 41° API |