Learn More
Thermo Scientific™ TRACE™ TR-5MS GC Columns
Product Code. p-4522510 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
10 m
100 m
15 m
20 m
30 m
60 m
Diameter (Metric) Inner:
0.1 mm
0.18 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.18 μm
0.25 μm
0.5 μm
1 μm
1.5 μm
3 μm
Unit Size:
Each
Product Code. 10239694
Brand: Thermo Scientific™ 260F020P
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Choose these colums that feature a popular GCMS phase for many applications.
Thermo Scientific™ TRACE TR-5MS GC Columns feature a popular GCMS phase for many applications.
  • Nonpolar phase, 5% phenyl polysilphenylene-siloxane
  • Low bleed and high stability
  • High signal-to-noise for increased sensitivity
  • High robustness to oxygen and water contamination
  • Similar to B-5, DB-5MS, DB-5.625 XTI-5, BPX5, Rtx-5MS, Rtx- 5, SilMS, AT-5, AT-5MS, 007-5MS, SPB-5, CP-Sil 8CB, Ultra-2, HP-5, HP-5MS, HP5-TA, SPB-5, MDN-5S, VF-5ms, RSL-200, CB-5, OV-5, PE-5, 007-2(MP-5), SE-52, SE-54, PTE-5, CC-5, RH-5ms, ZB-5
Recommended for:
  • Hydrocarbons
  • Solvents
  • Pesticides
  • Herbicides
  • Phenols
  • Amines
TRUSTED_SUSTAINABILITY
Specifications
Max. Temperature 360/370°C
Diameter (Metric) 0.1 mm
Diameter (Metric) Inner 0.1 mm
Product Line TRACE
Stationary Phase Trace™ TR-5 MS
USP Type G27, G36
For Use With (Application) Herbicides Analysis, GC/MS for Solvents, GC/MS for Pesticides, GC/MS for Herbicides, GC/MS for Phenols, GC/MS for Amines
Film Thickness 0.1 μm
Length (Metric) 10 m
Polarity Non-Polar
Description 10 m x 0.10 mm x 0.10 μm