Learn More
Thermo Scientific™ TRACE™ TR-225 GC Columns
Product Code. 10200155 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
15 m
30 m
60 m
Unit Size:
Each
Product Code. 10200155
Brand: Thermo Scientific™ 260Y130P
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Thermo Scientific™ TRACE TR-225 GC Columns offer reliable and reproducible performance.
Thermo Scientific™ TRACE TR-225 GC Columns offer reliable and reproducible performance.
  • Mid-polarity phase, 50% Cyanopropylphenyl polysiloxane
  • Low bleed even at elevated temperatures
  • Outstanding robustness for difficult separations
  • Manufactured to minimize risk of damage from contaminated carrier gas
  • Similar to: DB-225, HP-225, RTX-225, BP225
Recommended for:
  • Fatty Acid Methyl Esters (FAMEs)
  • Carbohydrates
  • Neutrol Sterols
TRUSTED_SUSTAINABILITY
Specifications
Max. Temperature 230/250°C
Diameter (Metric) 0.25 mm
Diameter (Metric) Inner 0.25 mm
Stationary Phase Trace™ TR-225
For Use With (Application) GC for Fatty Acid Methyl Esters (FAMEs), GC for Carbohydrates, GC for Neutral Sterols
Film Thickness 0.25 μm
Length (Metric) 15 m
Polarity Mid-Polar