Learn More
Kimberly-Clark™ Hostess™ NATURA™ Folded 1 Ply Paper Hand Towels 6019, 20 Packs
GREENER_CHOICE
Product Code. 10210289
Change view
Click to view available options
Color:
Green
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10210289 Green Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Product Code. 10210289 Supplier Kimberly-Clark™ Supplier No. 6019
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Sustainable green folded paper hand towels are manufactured in Germany with 100% recycled fibre. The Z fold paper towels are Blue Angel, FSC, and European Ecolabel certified for extra environmental assurance.
20 x 230 green, 1 ply sheets (4,600 total) One clip of folded paper towels contains 230 green, 1 ply disposable hand towels Green interfold paper towels manufactured in Germany with 100% recycled fibre The interfolded design of each paper towel ensures single sheet dispensing to reduce wastage and promote good washroom hygiene The Z fold paper hand towels are Blue Angel, FSC and European Ecolabel certified for extra environmental assurance Compatible with the Aquarius Folded hand towel dispenser in white (product code 6954) Provide a practical and consistent washroom experience with Hostess washroom solutions, designed for cost efficient quality in the workplace. Hostess NATURA folded hand towels are manufactured in Germany with 100% recycled fibre, the interfolded design of each paper towel ensures single sheet dispensing to reduce wastage, promote good hand hygiene and thereby boost workplace hygiene. Compatible with the Aquarius folded hand towel dispenser in white (product code 6954). The interfold hand towels are Blue Angel, FSC and European Ecolabel certified for extra environmental assurance. One clip of paper towels Z fold contains 230 green, 1 ply paper hand towels. Each Kimberly-Clark paper towel measures 25cm x 23cm. 20 x 230 green, 1 ply sheets (4,600 total)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Multi-fold Hand Towel
Length (English) 1.96 ft.
Length (Metric) 60 cm
Width (English) 1.41 ft.
Width (Metric) 43 cm
Material Paper
Color Green