Learn More
Brady™ ToughStripe Max Floor Corner L Marks
Product Code. 31900283
Change view
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
Dimensions (L x W):
127 x 50.8 mm
203.2 x 76.2 mm
254 x 101.6 mm
Unit Size:
Each
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Dimensions (L x W) unitSize
31900283 Yellow 127 x 50.8 mm Each
31900284 Blue 127 x 50.8 mm Each
31900282 Yellow 254 x 101.6 mm Each
31900261 White 127 x 50.8 mm Each
31900309 White 203.2 x 76.2 mm Each
31900299 Yellow 203.2 x 76.2 mm Each
31900303 Blue 203.2 x 76.2 mm Each
31900301 Green 127 x 50.8 mm Each
31900310 Black 203.2 x 76.2 mm Each
31900312 Black 127 x 50.8 mm Each
31900286 Green 203.2 x 76.2 mm Each
31900273 Red 127 x 50.8 mm Each
31900288 Black 254 x 101.6 mm Each
31900296 Blue 254 x 101.6 mm Each
31900253 Green 254 x 101.6 mm Each
31900266 Red 203.2 x 76.2 mm Each
31900308 Red 254 x 101.6 mm Each
31900257 White 254 x 101.6 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Product Code. 31900283 Supplier Brady™ Supplier No. 177039
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
Built stronger, lasts longer on punishing industrial floors. ToughStripe™ Max Floor Marking Tape is purpose-built to hold its ground in forklift and heavy traffic areas.
  • Re-engineered to be twice as thin yet twice as durable to better withstand damaging impacts in forklift and heavy traffic areas
  • Thinner profile and true edge-to-edge adhesive minimise pick points
  • Strong polyester (PETG) material is four times more rigid than vinyl and contains no plasticisers
  • Damage-isolating design keeps tape intact longer by preventing rips, gouges and tears from running
  • Matt finish provides excellent color contrast for clear visibility under any lighting
  • Compact construction results in a smaller, lightweight roll
  • Can be easily installed by one person with minimal tools
  • Compatible with the ToughStripe Floor Tape Applicator (part number 150852)
Specifications
Dimensions (L x W) 127 x 50.8 mm
Finish Gloss
For Use With (Application) Facility Identification, Floor Marking, Lean and 5S Labeling, Safety Identification, Warehouse Marking
Length (English) 5 in.
Length (Metric) 127 mm
Temperature Range (English) 50°F to 140°F
Temperature Range (Metric) 10°C to 60°C
Width (English) 2 in.
Width (Metric) 50.8 mm
Adhesive Type Rubber
Color Yellow
Material Polyester
Product Type Floor Marking Tape