Learn More
DiversifiedBiotech TOUGH-SPOTS™ Roll Labels
Click to view available options
Dimensions (LxW):
0.375 x 0.375 in.
0.50 x 0.50 in.
Unit Size:
Pack of 5000
Description
- Pre-cut peel off labels for tube tops with circular cryogenic
- Circular cryogenic labels formatted for handwriting
- Tough Spots™ on a roll are easy to write PVC cryogenic labels that accept writing from most lab markers.
- Tough Spots™ come in a variety of colors and sizes to ensure the right label for the right job
Specifications
Specifications
| Adhesive Type | Peel and Stick, Acrylic |
| Color | Assorted |
| Finish | Semi-gloss |
| Format | Roll |
| Material | Polyolefin |
| Print Technology | Laser |
| Product Type | Ultra Reflective Letter Label |
| Sterility | Non-sterile |
| Quantity | 5000/Pk. |
| Dimensions (LxW) | 0.375 x 0.375 in. |
| Show More |