Description
- Exceptional fit and feel: Made from high-quality elastic latex, TouchNTuff™ 69-318 gloves offer superior comfort
- Heightened sensitivity: The thin design provides excellent tactility when compared to standard latex gloves
- Secure handling: Textured fingers for easy handling of small objects and delicate instruments
- Food handling compliance: These gloves are food-handling compliant, and thus are suitable for a wide variety of food handling applications
Specifications
Specifications
| Applications | Industrial, Laboratory |
| Color | Natural |
| Disposable | Single-use |
| Exterior Finish | Fully Textured |
| Finger Thickness (English) | 5.5 mil |
| Finger Thickness (Metric) | 0.14 mm |
| Latex Free | No |
| Material | Natural Rubber Latex |
| Palm Thickness (English) | 4.7 mil |
| Palm Thickness (Metric) | 0.12 mm |
| Show More |