Learn More
WTW™ Total Nitrogen Test Kits
Click to view available options
Range:
0.5 to 15 mg/L
0.5 to 15 mg/L N Total
0.5 to 25 mg/L
10 to 150 mg/L
5 to 150 mg/L
Unit Size:
0.15kg
0.25kg
Each
Pack of 50
Description
- Variety of instrumentation for testing nitrogen in water
- For waste water, drinking water, surface water
- Nitrospectral Method
- Photometric Test Methods
- CombiCheck Total Nitrogen NTotal cell test kit
- Safe determinations of COD and NTotal in water
Specifications
Specifications
| Analytes | Chromosulfuric acid oxidation |
| For Use With (Application) | For waste water, drinking water, surface water |
| For Use With (Equipment) | For Photoflex™ photometers |
| No. of Tests | 30 |
| Range | 5 to 150 mg/L |
| Sufficient For | 25 Tests |
| Test Method | Photometric |
| Type | Combicheck COD Chemical oxygen demand O2 cell test kit |