missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Total Nitrogen Reagent Sets, For Colorimetric Water Testing

Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Total Nitrogen Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.

Brand:  Hanna Instruments™ HI93767B50

Additional Details : CAS : 7664-93-9 Weight : 0.15000kg Transport : UN number : 3316 Chem class : 9 Pack group : II


 View more versions of this product

Product Code. 11748763

  • 476.00€ / 0.15kg
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

These high quality reagents are designed to follow the chromotropic acid method for colorimetric determination of total nitrogen, in which a digestion is required to convert all forms of nitrogen to nitrate. Sodium metabisulfite is added to remove potential interferences. The reaction between nitrate and the reagents causes a yellow tint in the sample, and a compatible meter can be used to determine the concentration from the color that is produced.

  • Reagent sets are available for low range or high range determinations
  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability
Specifications

Specifications

Chromotropic Acid
Compatible Hanna benchtop and portable photometers
50
10 to150mg/L nitrogen
Water Testing Kit
Total nitrogen high range determination
Bottle and individual packets
1 Set
Cholorimetric determination
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • X50 TOTAL N REAGENT 106150 MG/L

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising solid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 A
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • EUH031-Contact with acids liberates toxic gas.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : potassium persulphate, sulphuric acid, sodium meta bisulphite

SDS
Documents

Documents

Certificates
Special Offers

Special Offers