Learn More
Hanna Instruments™ Total Nitrogen Reagent Sets, For Colorimetric Water Testing
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Total Nitrogen Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
476.00€ - 482.00€
Specifications
Analytes | Chromotropic Acid |
---|---|
For Use With (Equipment) | Compatible Hanna benchtop and portable photometers |
Includes | Bottle and individual packets |
No. of Tests | 50 |
Quantity | 1 Set |
Product Code | Brand | For Use With (Application) | No. of Tests | Range | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | No. of Tests | Range | Price | Quantity & Availability | |||
11738763
|
Hanna Instruments™
HI93767A50 |
Total nitrogen low range determination | 50 | 0.0 to 25.0mg/L nitrogen |
482.00€
Pack of 50 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||
11748763
|
Hanna Instruments™
HI93767B50 |
Total nitrogen high range determination | 50 | 10 to150mg/L nitrogen |
476.00€
0.15kg |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||
Description
These high quality reagents are designed to follow the chromotropic acid method for colorimetric determination of total nitrogen, in which a digestion is required to convert all forms of nitrogen to nitrate. Sodium metabisulfite is added to remove potential interferences. The reaction between nitrate and the reagents causes a yellow tint in the sample, and a compatible meter can be used to determine the concentration from the color that is produced.
- Reagent sets are available for low range or high range determinations
- Pre-made, pre-measured water testing reagents for ease of use
- Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
- Clearly marked with expiration date and lot number on each packet and bottle for traceability
Specifications
Chromotropic Acid | |
Bottle and individual packets | |
1 Set | |
Water Testing Kit |
Compatible Hanna benchtop and portable photometers | |
50 | |
Cholorimetric determination |