Learn More
Lovibond™ Total Hardness tablets
Product Code. 15381995
Change view
Click to view available options
Quantity:
250 Tablets
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15381995 250 Tablets Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15381995 Supplier Lovibond™ Supplier No. 503551
  • ONLINE EXCLUSIVE PRICE
    19.00€
    15.70€ / Pack of 250
    Save 3.30€
    17% Off
View all promotions
 Restricted Item
Estimated Shipment: 10-12-2026
The suitable tablet for the fast result
Water analysis couldn’t be easier. Precise dosing, long shelf life and easy handling make the tablet reagent the most popular indicator system and the cheapest way to have analyse a parameter.
Total Hardness tablets 250pcs in a bottle.  
Specifications
Format Tablet
Quantity 250 Tablets
Type Reagent
compliance-icons compliance-icons
Product Identifier
  • X250 TOTAL HARDNESS TABLETS
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: lithium hydroxide