Learn More
Hanna Instruments™ Hardness Reagent Sets, For Colorimetric Water Testing
Product Code. 11793363
Click to view available options
Quantity:
1 Set
Unit Size:
Pack of 100
Product Code. 11793363
Brand: Hanna Instruments™ HI9373502
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Hardness Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
These high quality reagents are designed to follow accepted test methods for colorimetric determination of various parameters of water hardness. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
  • Reagent sets are available for determination of:
    • Calcium Hardness (using liquid or powdered reagents)
    • Calcium and Magnesium Hardness (high range)
    • Magnesium and Total Hardness
    • Total Hardness (low, medium and/or high range)
  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability
Specifications
For Use With (Application) Total hardness high range determination
For Use With (Equipment) Compatible Hanna benchtop and portable photometers
Includes 2 dropper bottles and 100 individual packets
No. of Tests 100
Quantity 1 Set
Range 400 to 750mg/L as CaCO3 total hardness
Test Method EPA recommended method 130.1
Type Water Testing Kit
compliance-icons compliance-icons compliance-icons
Product Identifier
  • X100 HARDNESS REAGENT 4006750 MG/
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contains : Tetrasodium EDTA, Propionic acid, 2-Amino-2-methyl-1-propanol