missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Total Acid Number Standards, Paragon Scientific™
Click to view available options
Concentration:
0.10 mg KOH/g
0.49 mg KOH/g
0.99 mg KOH/g
1.54 mg KOH/g
2.02 mg KOH/g
2.48 mg KOH/g
2.93 mg KOH/g
4.57 mg KOH/g
Quantity:
50 g
125 g
3 x 50 g
Description
Each standard is certified in strict accordance with international test method protocols, ASTM D664 / IP 177. Manufacture and certification is carried out in strict accordance with Paragon Scientific’s dual UKAS accreditations to ISO 17025 & ISO 17034. This dual accreditation allows us to ensure the highest level of accreditation guarantee and provides the most credible certified data available worldwide.
- Acid Number (TAN) tested and certified in accordance with ASTM D664 / IP 177
- Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation
- Highest level of accreditation guarantee, providing the most credible certified data available worldwide
- Fully traceable to international standards and low levels of uncertainty achieves maximum accuracy of data
- Typically used in, but not limited to, the analysis of used oils and lubricants
- Supplied as single volume packs or as triple packs
- Supplied in tamper-evident glass packaging, providing assurance of sample integrity
- Manufactured in the United Kingdom
- 24 month shelf life
Specifications
Concentration | 1.54 mg KOH/g |
Quantity | 125 g |
Safety and Handling
Product Identifier
- TAN015
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Nominal values quoted
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'