missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Toremifene, 98+%
A chlorinated tamoxifen analogue that induces apoptosis in some cells
110.00€ - 182.00€
Chemical Identifiers
| CAS | 89778-26-7 |
|---|---|
| Formula Weight | 405.96g/mol |
| Synonym | Toremifene base; 2-(4-[(1Z)-4-Chloro-1,2-diphenyl-1-butenyl]phenoxy)-N,N-dimethylethanamine |
| Format | Powder |
| For Use With (Application) | A chlorinated tamoxifen analogue that induces apoptosis in some cells |
| Description | White to Off-white |
| Melting Point | 108°C to 110°C |
| Product Type | Apoptosis Reagent |
| Assay | ≥98% |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15425979
|
Thermo Scientific™
J63803.ME |
500 mg |
110.00€
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15405979
|
Thermo Scientific™
J63803.03 |
1 g |
182.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 89778-26-7 | |
| Toremifene base; 2-(4-[(1Z)-4-Chloro-1,2-diphenyl-1-butenyl]phenoxy)-N,N-dimethylethanamine | |
| A chlorinated tamoxifen analogue that induces apoptosis in some cells | |
| 108°C to 110°C | |
| ≥98% |
| 405.96g/mol | |
| Powder | |
| White to Off-white | |
| Apoptosis Reagent |
Specifications
| White to Off-white | |
| 89778-26-7 | |
| 405.96g/mol | |
| 108°C to 110°C | |
| A chlorinated tamoxifen analogue that induces apoptosis in some cells |
| Powder | |
| C26H28ClNO | |
| Toremifene base; 2-(4-[(1Z)-4-Chloro-1,2-diphenyl-1-butenyl]phenoxy)-N,N-dimethylethanamine | |
| Apoptosis Reagent | |
| ≥98% |
Safety and Handling
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title