Learn More
Thermo Scientific™ TopVision Low Melting Point Agarose Promotion
Product Code. 15707546 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15707546 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15707546 Supplier Thermo Scientific™ Supplier No. R0801
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Perform in-gel enzymatic process experiments and nucleic acid recovery after electrophoresis with Thermo Scientific™ TopVision™ Low Melting Point Agarose.
Top Vision Low Melting Point Agarose is highly purified agarose with a low melting temperature certified by strict quality control test procedures. It is used for in-gel enzymatic process experiments and for nucleic acids recovery from gels using Agarase (#EO0461).
Highlights
  • Optimal concentration between 0.7-2% in all typical buffer systems
  • GQ (Genetic Quality) certified, which ensures that nucleic acids recovered from preparative gels can be used for downstream applications (enzymatic reactions etc.)
  • Low DNA/RNA binding 
  • Excellent gel transparency 
  • DNase and RNase free 
  • Suitable for RNA analysis 
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 g
Product Type Agarose Gel