Learn More
Fisherbrand™ Top-Line Pipette Tips
Product Code. p-8002104 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.5 to 20 μL
1 to 200 μL
5 to 300 μL
Sterility:
Non-sterile
Sterile
Color:
Clear
Yellow
Length (Metric):
34.1mm
45.7mm
50.8mm
55mm
Unit Size:
Pack of 1000
Pack of 960
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Color Length (Metric) unitSize
10182742 0.1 to 10 μL Non-sterile Clear 34.1mm Pack of 960
10346052 0.5 to 20 μL Non-sterile Clear 45.7mm Pack of 1000
10739444 1 to 200 μL Non-sterile Clear 50.8mm Pack of 960
10739254 1 to 200 μL Non-sterile Yellow 50.8mm Pack of 1000
10233932 0.1 to 10 μL Sterile Clear 34.1mm Pack of 960
10458072 1 to 200 μL Sterile Clear 50.8mm Pack of 960
10142172 1 to 200 μL Sterile Yellow 50.8mm Pack of 960
10532543 0.5 to 20 μL Non-sterile Clear 45.7mm Pack of 960
10542543 0.5 to 20 μL Sterile Clear 45.7mm Pack of 960
10509862 5 to 300 μL Non-sterile Clear 55mm Pack of 1000
10396142 5 to 300 μL Non-sterile Clear 55mm Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 10182742
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Eliminate sample contamination and prevent sample loss. . Fisherbrand™ Top-Line Pipette Tips provide precision pipetting with conical tip and secure fit.
  • Optimized cone geometry for a secure fit
  • Graduation marks on tip at standard volumes
  • Conical tip opening prevents drop formation at the end of the tip
  • Tips made of autoclavable, non-wettable, virgin polypropylene
  • Sterile packs available (beta irradiation)
Specifications
Volume (Metric) 0.1 to 10 μL
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Length (Metric) 34.1mm
Certifications/Compliance ISO Standard 8655, EU-Directive 98/79/EG, competent authority code DE/CA90, EN 552/ISO 11137 and DIN EN 556, EU Directive 2002/95/EC, ISO Standard 14000 and ISO Standard 14001, ISO Standard 10993, DIN Standard 12600
Sterilization Method Autoclave
Format Rack
Tip Style Universal
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line Top-Line
Material Virgin Polypropylene
Graduations Standard volumes