Learn More
Fisherbrand™ Top-Line Pipette Tips
Product Code. 10396142 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.5 to 20 μL
1 to 200 μL
5 to 300 μL
Sterility:
Non-sterile
Sterile
Color:
Clear
Yellow
Length (Metric):
34.1mm
45.7mm
50.8mm
55mm
Unit Size:
Pack of 1000
Pack of 960
Product Code. 10396142
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Eliminate sample contamination and prevent sample loss. . Fisherbrand™ Top-Line Pipette Tips provide precision pipetting with conical tip and secure fit.
  • Optimized cone geometry for a secure fit
  • Graduation marks on tip at standard volumes
  • Conical tip opening prevents drop formation at the end of the tip
  • Tips made of autoclavable, non-wettable, virgin polypropylene
  • Sterile packs available (beta irradiation)
Specifications
Volume (Metric) 5 to 300 μL
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Length (Metric) 55mm
Certifications/Compliance ISO Standard 8655, EU-Directive 98/79/EG, competent authority code DE/CA90, EN 552/ISO 11137 and DIN EN 556, EU Directive 2002/95/EC, ISO Standard 14000 and ISO Standard 14001, ISO Standard 10993, DIN Standard 12600
Sterilization Method Autoclave
Format Rack
Tip Style Universal
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line Top-Line
Material Virgin Polypropylene
Graduations Standard volumes