Learn More
(±)-Tonalide-d11 (5,8-d2; 4,4,7-trimethyl-d9), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Tonalide-D11 (5,8-D2,4,7,7-trimethyl-D9) |
| CAS | 2714413-99-5 |
| Molecular Formula | C18 D11 H15 O |
| Quantity | 0.005 g |
| Synonym | 1-[5,6,7,8-Tetrahydro-5,5,6-trimethyl-3,8,8-tri(methyl-d3)-2-naphthalenyl-1,4-d2]ethanone, 6-Acetyl-1,1,2-trimethyl-4,4,7-tri(methyl-d3)tetraline-5,8-d2, Tonalide-d11, AHTN-d11, Tonalide-D11 (5,8-D2,4,7,7-trimethyl-D9) |
| InChI Formula | InChI=1 S/C18H26O/c1-11-8-16-15(9-14(11)13(3)19)17(4,5)10-12(2)18(16,6)7/h8-9,12 H,10H2,1-7H3/i1D3,4D3,5D3,8 D,9 D |
| SMILES | [2 H]c1c(C(=O)C)c(c([2 H])c2c1C(CC(C)C2(C)C)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1-[1,4-dideuterio-5,5,6-trimethyl-3,8,8-tris(trideuteriomethyl)-6,7-dihydronaphthalen-2-yl]ethanone |
| Molecular Weight (g/mol) | 269.4662 |
| Formula Weight | 269.2674 g/mol |
| Show More |