Learn More
Tolazoline, TRC
Specifications
Specifications
| Chemical Name or Material | Tolazoline |
| CAS | 59-98-3 |
| Molecular Formula | C10H12N2 |
| Quantity | 5 g |
| Synonym | 2-Imidazoline','2-benzyl- (8CI), Imidazoline','2-benzyl- (6CI), 4,5-Dihydro-2-(phenylmethyl)-1H-imidazole, 2-Benzyl-2-imidazoline, 2-Benzyl-4,5-dihydro-1H-imidazole, 2-Benzyl-4,5-imidazoline, 2-Benzylimidazoline, Artonil, Benzazoline, Benzidazol, Benzylimidazoline, Dilatol ASI, Divascol, Imidalin, Imidaline, Kasimid, Lambril, NSC 35110, Olitensol, Peripherine, Phenylmethylimidazoline, Prefaxil, Priscol, Priscoline, Vasimid, Vasodil |
| InChI Formula | InChI=1S/C10H12N2/c1-2-4-9(5-3-1)8-10-11-6-7-12-10/h1-5H,6-8H2,(H,11,12) |
| SMILES | C(C1=NCCN1)c2ccccc2 |
| IUPAC Name | 2-benzyl-4,5-dihydro-1H-imidazole |
| Molecular Weight (g/mol) | 160.22 |
| Formula Weight | 160.1 |