Learn More
Tocris Bioscience™ SNX 482
Product Code. 18015152
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. 18015152
Brand: Tocris Bioscience™ 2945/10U
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Potent and selective CaV2.3 blocker
SNX 482 is a potent and selective, voltage-dependent R-type CaV2.3 calcium channel blocker (IC50 = 30 nM). Antinociceptive; inhibits nociceptive C-fibre and Aδ-fibre-evoked neuronal responses.