Learn More
Tocris Bioscience™ SNX 482
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
SNX 482 is a potent and selective, voltage-dependent R-type CaV2.3 calcium channel blocker (IC50 = 30 nM). Antinociceptive; inhibits nociceptive C-fibre and Aδ-fibre-evoked neuronal responses.
Specifications
Specifications
| Quantity | 10 μg |