Learn More
Tocris Bioscience™ Neurokinin B (human, porcine)
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Neurokinin B (human, porcine) is an endogenous tachykinin agonist peptide that shows preference for the NK3 receptor (EC50 = 1 nM).
Specifications
Specifications
| Purity | 95% |
| Content And Storage | Store at -20°C |
| Quantity | 1 mg |