Learn More
R&D Systems™ Leupeptin hemisulfate
Description
Leupeptin hemisulfate is a reversible inhibitor of trypsin-like and cysteine proteases such as calpain. Shown to inhibit activation-induced programmed cell death.
Specifications
Specifications
| Product Line | Tocris |
| Purity | 0.95 |
| Content And Storage | Store at -20°C |
| Quantity | 25 mg |