Learn More
Tocris Bioscience™ N-Formyl-Met-Leu-Phe
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
N-Formyl-Met-Leu-Phe is an endogenous chemotactic peptide and agonist for the formyl peptide receptor 1 (FPR1) (Ki = 38 nM). Stimulates aggregation of leukocytes.
Specifications
Specifications
| Purity | 0.95 |
| Content And Storage | Store at -20°C |
| Quantity | 1 mg |