missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Ghrelin (rat)

Product Code. 18074482
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18074482 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18074482 Supplier R&D Systems™ Supplier No. 1465/1
  • ONLINE EXCLUSIVE PRICE
    738.06€
    659.40€ / 1mg
    Save 78.66€
    10% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Endogenous ghrelin receptor agonist

Ghrelin (rat) is an endogenous agonist peptide for the ghrelin receptor (GHS-R1a). Produced mainly by the stomach, Ghrelinstimulates release of growth hormone from the pituitary gland in vitro and in vivo, and regulates feeding, growth and energy production.
TRUSTED_SUSTAINABILITY

Specifications

Product Line Tocris
Purity 0.95
Content And Storage Store at -20°C
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.