missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ghrelin (human)

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Ghrelin (human) is an endogenous agonist peptide for the ghrelin receptor (GHS-R1a). Produced mainly by the stomach, it stimulates release of growth hormone from the pituitary gland in vitro and in vivo, and regulates feeding, growth and energy production.
Specifications
Specifications
| Purity | 0.95 |
| Content And Storage | Store at -20°C |
| Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction