Learn More
Tocris Bioscience™ Gastrin I (human)
GREENER_CHOICE
Product Code. 18094612
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 18094612
Brand: Tocris Bioscience™ 3006/1
Selective CCK2 agonist
Gastrin I (human) is an endogenous peptide produced in the stomach that acts as a selective CCK2 receptor agonist. Gastrin I stimulates gastric acid secretion and has a mitogenic effect on gastric cells. Gastrin stimulates gastric epithelial cell proliferation and histamine secretion (EC50 values are 6.2 pM and 0.014 nM, respectively). Gastrin I is also used in the culture of stomach, intestine and pancreas organoids. For more information about using Gastrin I in the generation of gastric organoids see the Human Gastric Organoid Culture Protocol .
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 mg