Learn More
Tocris Bioscience™ [D-Lys3]-GHRP-6
Ghrelin receptor antagonist
Brand:  Tocris Bioscience™ 1922/5
Encompass
Product Code. 13480234
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
[D-Lys3]-GHRP-6 is an antagonist at the ghrelin receptor (GHS-R1a) (IC50 = 0.9 μM). Also weakly binds to melanocortin receptors (Ki = 26-120 μM). Centrally active in vivo.
Specifications
Specifications
5 mg