Learn More
Tocris Bioscience™ Cyclotraxin B
TrkB receptor antagonist
Brand: Tocris Bioscience™ 5062/1
This item is not returnable.
View return policy
Description
Cyclotraxin B is an antagonist of TrkB receptors; inhibits BDNF-induced TrkB activity (IC50 = 0.30 nM). Allosterically alters TrkB receptor conformation but does not alter BDNF binding. Prevents BDNF-induced cold allodynia in mice. Also shown to exhibit putative anxiolytic properties in mice.Specifications
| 1 mg |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.