Learn More
Tocris Bioscience™ Cyclotraxin B
GREENER_CHOICE
TrkB receptor antagonist
Brand:  Tocris Bioscience™ 5062/1
Encompass
Product Code. 15726426
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
Cyclotraxin B is an antagonist of TrkB receptors; inhibits BDNF-induced TrkB activity (IC50 = 0.30 nM). Allosterically alters TrkB receptor conformation but does not alter BDNF binding. Prevents BDNF-induced cold allodynia in mice. Also shown to exhibit putative anxiolytic properties in mice.