Learn More
Tocris Bioscience™ Compstatin
GREENER_CHOICE
Product Code. 18014102
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 18014102
Brand: Tocris Bioscience™ 2585/1
C3-binding protein, inhibits complement activation
Compstatin is a complement inhibitor; binds C3 inhibiting proteolytic cleavage by C3 convertase (IC50 = 28 μM) and activation of classical and alternative complement pathways (IC50 values are 63 and 12 μM respectively). Prolongs graft survival and reduces hyperacute rejection of ex vivo pig kidneys perfused with human blood and inhibits in vivo heparin/protamine-induced complement activation in primates.
Control Peptide also available.
TRUSTED_SUSTAINABILITY
Specifications
Purity 0.95
Quantity 1 mg