missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BQ-3020

Click to view available options
Quantity:
500 μg
Unit Size:
500µg
Description
BQ-3020 is a highly potent and selective ETB endothelin receptor agonist (Ki values are 0.18 and 970 nM at human ETB and ETA receptors respectively).
Specifications
Specifications
| Purity | 0.95 |
| Content And Storage | Store at -20°C |
| Quantity | 500 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction