Learn More
Tocris Bioscience™ BDNF (human)
GREENER_CHOICE
Product Code. 18004612
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. 18004612
Brand: Tocris Bioscience™ 2837/10U
Activates TrkB and p75 receptors
BDNF (human) is a member of the neurotrophin growth factor family that binds and activates TrkB and p75 neurotrophin receptors. BDNF enhances the survival, growth and differentiation of neurons. BDNF expression is altered in neurodegenerative disorders such as Parkinson's and Alzheimer's disease. In an animal model of Huntington's disease, BDNF administration or overexpression rescues the disease phenotype.
TRUSTED_SUSTAINABILITY
Specifications
Purity 0.95
Quantity 10 μg