missing translation for 'onlineSavingsMsg'
Learn More

Melford Tobramycin Sulfate

Product Code. 30018862
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 30018862

Brand: Melford T450001.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tobramycin Sulfate is an aminoglycoside antibiotic that inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, leading to the misreading of mRNA and preventing the proper initiation complex formation.

Tobramycin Sulfate is an aminoglycoside antibiotic that inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, leading to the misreading of mRNA and preventing the proper initiation complex formation. This interference disrupts the translation process and inhibits bacterial growth.

Tobramycin is used in microbiology for selective bacterial isolation and in antibiotic sensitivity testing to determine the susceptibility of bacterial strains. Ideal for studying bacterial susceptibility patterns and antibiotic resistance. Its inclusion in culture media can help selectively isolate bacteria that are susceptible to this antibiotic.

Specifications

Chemical Name or Material Tobramycin
CAS 79645-27-5, 49842-07-1
Quantity 1 g
Molecular Formula (C18H37N5O9)2-5H2SO4
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.