missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF Rat anti-Mouse, BB700, Clone: MP6-XT22, BD Horizon™
Rat Monoclonal Antibody
272.00€ - 645.00€
Specifications
Antigen | TNF |
---|---|
Clone | MP6-XT22 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15815119
|
BD Biosciences
566511 |
25 μg |
272.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
15805119
|
BD Biosciences
566510 |
0.1 mg |
645.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
Description
TNF Monoclonal antibody specifically detects TNF in Mouse samples. It is validated for Flow Cytometry.Specifications
TNF | |
0.2 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
IgG1 | |
Refrigerate (2 to 8°C) |
MP6-XT22 | |
Flow Cytometry | |
Brilliant Blue 700 (BB700) | |
Rat | |
Mouse | |
Primary |