Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| Target | TMPRSS8 |
| Synonym | Disp, FLJ16659, MGC52282, protease, serine, 30 homolog (mouse), pseudogene, TMPRSS8, TMPRSS8P |
| Gene Symbol | PRSS30P |