missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TMEM41B Antibody Blocking Peptide

Product Code. 18639946 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
Product Code. Quantity unitSize
18639946 0.1 mg 0.10mg
1 options
This item is not returnable. View return policy

Product Code. 18639946

Brand: Novus Biologicals™ NBP300527PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Blocking Peptide

TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 440026
Concentration 1 mg/mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP3-00527.
Gene Alias KIAA0033MGC33897, MGC57262, transmembrane protein 41 B
Gene Symbol TMEM41B
Quantity 0.1 mg
Research Category Autophagy, Lipid and Metabolism
Storage Buffer Peptide dissolved in dH20. Contains no BSA.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.