missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TMEM23 Recombinant Protein

Brand:  Abnova™ H00259230-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16159763

  • 545.00€ / 25µg

Estimated Shipment: 22-08-2022
to see stock.
Add to basket

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH42899
Solution
259230
TMEM23 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SGMS1
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
71.17
8
Glutathione Sepharose 4 Fast Flow
25 ug
MKEVVYWSPKKVADWLLENAMPEYCEPLEHFTGQDLINLTQEDFKKPPLCRVSSDNGQRLLDMIETLKMEHHLEAHKNGHANGHLNIGVDIPTPDGSFSIKIKPNGMPNGYRKEMIKIPMPELERSQYPMEWGKTFLAFLYALSCFVLTTVMISVVHERVPPKEVQPPLPDTFFDHFNRVQWAFSICEINGMILVGLWLIQWLLLKYKSIISRRFFCIVGTLYLYRCITMYVTTLPVPGMHFNCSPKLFGDWEAQLRRIMKLIAGGGLSITGSHNMCGDYLYSGHTVMLTLTYLFIKEYSPRRLWWYHWICWLLSVVGIFCILLAHDHYTVDVVVAYYITTRLFWWYHTMANQQVLKEASQMNLLARVWWYRPFQYFEKNVQGIVPRSYHWPFPWPVVHLSRQVKYSRLVNDT
MGC17342/MOB/MOB1/SMS1/TMEM23
SGMS1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers