missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM16A/DOG1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Specifications
Antigen | TMEM16A/DOG1 |
---|---|
Concentration | 0.24 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16870484
|
Proteintech
12652-1-AP-20UL |
20 μL |
140.00€
20µL |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
16860484
|
Proteintech
12652-1-AP-150UL |
150 μL |
399.00€
150µL |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
Description
This antibody can recognize isoform1 and isoform2 of human ANO1.
DOG-1, the novel marker, is expressed ubiquitously in gastrointestinal stromal tumors irrespective of kit or PDGFR alpha mutation status.Specifications
TMEM16A/DOG1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Human | |
Q5XXA6 | |
309135, 55107 | |
TMEM16A/DOG1 Fusion Protein Ag3320 | |
Primary | |
-20°C | |
ANO1 |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ANO1, Anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A, TMEM16A/DOG1, Transmembrane protein 16A | |
Ano1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |